Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdh-4
Entrez Gene ID
175590
Gene Name
Cadherin-4
Gene Aliases
CELE_F25F2.2
Chromosome Location
Chr.III: 4520357 - 4536994 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330837.3 NP_001317871.1 12 - 13 5365 101

View Full Product Details