Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-1
Entrez Gene ID
191828
Gene Name
Serpentine receptor class gamma-1
Gene Aliases
CELE_C18F10.4
Chromosome Location
Chr.III: 6256948 - 6258362 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065965.3 NP_498366.2 3 - 4 752 83

View Full Product Details