Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-13
Entrez Gene ID
176083
Gene Name
Zinc finger protein lin-13
Gene Aliases
CELE_C03B8.4
Chromosome Location
Chr.III: 7700338 - 7709495 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066277.5 NP_498678.3 19 - 20 6271 65
GenBank mRNA AF245435.1 - 19 - 20 6248 65

View Full Product Details