Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
such-1
Entrez Gene ID
176583
Gene Name
Suppressor of spindle checkpoint defect 1
Gene Aliases
CELE_Y66D12A.17
Chromosome Location
Chr.III: 11512850 - 11523759 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067084.6 NP_499485.2 7 - 8 1344 125

View Full Product Details