Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-84
Entrez Gene ID
176601
Gene Name
Galectin
Gene Aliases
CELE_Y41C4A.5
Chromosome Location
Chr.III: 11700259 - 11702862 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067113.7 NP_499514.3 4 - 5 385 107

View Full Product Details