Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- cap-1
- Entrez Gene ID
-
177497
- Gene Name
- F-actin-capping protein subunit alpha
- Gene Aliases
-
CELE_D2024.6
- Chromosome Location
-
Chr.IV: 7228403 - 7230223 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_068744.5 |
NP_501145.1 |
2 - 3 |
203 |
83 |
GenBank mRNA |
Z18853.1 |
- |
- |
211 |
83 |
- Gene Symbol
- D2024.5
- Entrez Gene ID
-
177496
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_D2024.5
- Chromosome Location
-
Chr.IV: 7227275 - 7230210 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383116.1 |
|
2 - 3 |
190 |
83 |
View Full Product Details