Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lys-10
Entrez Gene ID
184622
Gene Name
LYSozyme
Gene Aliases
CELE_F17E9.11
Chromosome Location
Chr.IV: 8332826 - 8333518 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069004.3 NP_501405.1 1 - 1 642 75

View Full Product Details