Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-15
Entrez Gene ID
177770
Gene Name
protein-coding
Gene Aliases
CELE_C10C5.6
Chromosome Location
Chr.IV: 9386208 - 9394992 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306928.3 NP_001293857.1 16 - 17 4677 96
NM_001306929.3 NP_001293858.1 16 - 17 4702 96
NM_001380409.1 NP_001368188.1 17 - 18 4733 96
NM_001380410.1 NP_001366749.1 17 - 18 4733 96
NM_069252.7 NP_501653.1 16 - 17 4693 96
GenBank mRNA AY396716.1 - 17 - 18 4733 96

View Full Product Details