Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-15
Entrez Gene ID
177770
Gene Name
protein-coding
Gene Aliases
CELE_C10C5.6
Chromosome Location
Chr.IV: 9386208 - 9394992 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380409.1 NP_001368188.1 4 - 5 461 76
NM_001380410.1 NP_001366749.1 4 - 5 461 76
NM_069252.7 NP_501653.1 3 - 4 403 76
GenBank mRNA AY396716.1 - 4 - 5 461 76

View Full Product Details