Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
egl-38
Entrez Gene ID
177876
Gene Name
Paired domain-containing protein
Gene Aliases
CELE_C04G2.7
Chromosome Location
Chr.IV: 10106599 - 10110517 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069435.6 NP_501836.1 1 - 2 161 63
GenBank mRNA AF002702.1 - 1 - 2 208 63

View Full Product Details