Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gta-1
Entrez Gene ID
177897
Gene Name
protein-coding
Gene Aliases
CELE_K04D7.3
Chromosome Location
Chr.IV: 10180660 - 10183939 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069461.4 NP_501862.1 4 - 5 317 129

View Full Product Details