Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ubxn-2
Entrez Gene ID
177057
Gene Name
protein-coding
Gene Aliases
CELE_Y94H6A.9
Chromosome Location
Chr.IV: 2720313 - 2725819 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028419.5 NP_001023590.1 1 - 2 129 58
NM_001372754.2 NP_001360209.1 2 - 2 129 58

View Full Product Details