Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-83
Entrez Gene ID
177065
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_Y54G2A.14
Chromosome Location
Chr.IV: 2839371 - 2841472 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067859.10 NP_500260.3 4 - 5 589 73

View Full Product Details