Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-74
Entrez Gene ID
189948
Gene Name
C-type lectin domain-containing protein
Gene Aliases
CELE_Y46C8AL.8
Chromosome Location
Chr.IV: 3957005 - 3960185 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068045.3 NP_500446.2 2 - 3 518 97

View Full Product Details