Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-33
Entrez Gene ID
177191
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_F36A4.6
Chromosome Location
Chr.IV: 4263293 - 4265223 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068123.5 NP_500524.2 3 - 4 217 71

View Full Product Details