Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exos-2
Entrez Gene ID
177403
Gene Name
EXOSome (multiexonuclease complex) component
Gene Aliases
CELE_Y73B6BL.3
Chromosome Location
Chr.IV: 6415478 - 6417460 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068577.4 NP_500978.1 3 - 4 564 122

View Full Product Details