Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
phy-2
Entrez Gene ID
178170
Gene Name
Prolyl 4-hydroxylase subunit alpha-2
Gene Aliases
CELE_F35G2.4
Chromosome Location
Chr.IV: 12211098 - 12215987 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069916.6 NP_502317.1 6 - 7 1124 131

View Full Product Details