Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dct-19
Entrez Gene ID
183136
Gene Name
CUB_2 domain-containing protein
Gene Aliases
CELE_C32H11.13
Chromosome Location
Chr.IV: 12942933 - 12944526 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070074.4 NP_502475.2 7 - 8 920 68

View Full Product Details