Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lev-1
Entrez Gene ID
178269
Gene Name
protein-coding
Gene Aliases
CELE_F09E8.7
Chromosome Location
Chr.IV: 13174157 - 13179457 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268775.4 NP_001255704.1 6 - 7 1291 119
NM_001268776.2 NP_001255705.1 5 - 6 1503 119
GenBank mRNA X98601.1 - - 1416 119

View Full Product Details