Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nol-9
Entrez Gene ID
187190
Gene Name
protein-coding
Gene Aliases
CELE_K09B11.2
Chromosome Location
Chr.IV: 13422500 - 13429399 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268815.2 NP_001255744.1 4 - 5 533 86

View Full Product Details