Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nol-9
Entrez Gene ID
187190
Gene Name
protein-coding
Gene Aliases
CELE_K09B11.2
Chromosome Location
Chr.IV: 13422500 - 13429399 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268815.2 NP_001255744.1 9 - 10 1532 100
NM_001268816.3 NP_001255745.1 1 - 2 258 100

View Full Product Details