Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fcd-2
Entrez Gene ID
178418
Gene Name
protein-coding
Gene Aliases
CELE_Y41E3.9
Chromosome Location
Chr.IV: 15025706 - 15037036 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268919.2 NP_001255848.1 9 - 10 4143 80
NM_001268920.3 NP_001255849.1 5 - 6 2364 80
NM_001268921.3 NP_001255850.1 3 - 4 1293 80

View Full Product Details