Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-42
Entrez Gene ID
190670
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_Y73F8A.3
Chromosome Location
Chr.IV: 15250152 - 15252257 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070439.4 NP_502840.4 3 - 4 658 150

View Full Product Details