Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srt-52
Entrez Gene ID
189650
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_Y38C9B.2
Chromosome Location
Chr.V: 49956 - 56046 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070728.2 NP_503129.2 2 - 3 348 65

View Full Product Details