Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ruvb-2
Entrez Gene ID
177458
Gene Name
RuvB-like 2
Gene Aliases
CELE_T22D1.10
Chromosome Location
Chr.IV: 6917107 - 6920192 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068666.5 NP_501067.1 2 - 3 149 122

View Full Product Details