Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abt-4
Entrez Gene ID
178559
Gene Name
ABC Transporter family
Gene Aliases
CELE_Y39D8C.1
Chromosome Location
Chr.V: 323772 - 330725 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070774.4 NP_503175.1 12 - 13 3964 74

View Full Product Details