Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-58
Entrez Gene ID
178573
Gene Name
Nuclear Hormone Receptor family
Gene Aliases
CELE_R11G11.2
Chromosome Location
Chr.V: 532259 - 534070 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070835.6 NP_503236.1 5 - 6 900 82

View Full Product Details