Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp-32B1
Entrez Gene ID
178697
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_Y5H2B.5
Chromosome Location
Chr.V: 2197997 - 2203462 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071197.8 NP_503598.1 6 - 7 1704 118

View Full Product Details