Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-82
Entrez Gene ID
191989
Gene Name
Seven TM Receptor
Gene Aliases
CELE_B0213.8
Chromosome Location
Chr.V: 3974661 - 3976264 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071704.2 NP_504105.2 2 - 3 269 113

View Full Product Details