Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cwp-1
Entrez Gene ID
178876
Gene Name
Coexpressed With Polycystins
Gene Aliases
CELE_C37H5.10
Chromosome Location
Chr.V: 4840617 - 4841279 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071893.5 NP_504294.1 2 - 3 170 74

View Full Product Details