Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-32
Entrez Gene ID
190384
Gene Name
GLOBIN domain-containing protein
Gene Aliases
CELE_Y58A7A.6
Chromosome Location
Chr.V: 5087508 - 5089746 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071962.5 NP_504363.3 2 - 3 296 77

View Full Product Details