Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-66
Entrez Gene ID
188119
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T05C3.7
Chromosome Location
Chr.V: 5480750 - 5482415 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072048.2 NP_504449.2 4 - 5 830 149

View Full Product Details