Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ubh-3
- Entrez Gene ID
-
259626
- Gene Name
- Ubiquitin carboxyl-terminal hydrolase
- Gene Aliases
-
CELE_Y40G12A.1
- Chromosome Location
-
Chr.V: 6506754 - 6509817 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072252.7 |
NP_504653.2 |
5 - 6 |
1299 |
71 |
- Gene Symbol
- W01A11.7
- Entrez Gene ID
-
189079
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_W01A11.7
- Chromosome Location
-
Chr.V: 6504540 - 6508805 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_072251.6 |
NP_504652.2 |
2 - 3 |
429 |
71 |
View Full Product Details