Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lbp-4
Entrez Gene ID
179161
Gene Name
Fatty acid-binding protein homolog 4
Gene Aliases
CELE_ZK742.5
Chromosome Location
Chr.V: 7820578 - 7821477 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072615.5 NP_505016.1 1 - 2 168 104

View Full Product Details