Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-50
Entrez Gene ID
179284
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_T27E4.6
Chromosome Location
Chr.V: 9069857 - 9073540 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322580.2 NP_001309487.1 6 - 7 1019 141
NR_136394.1 5 - 6 820 141

View Full Product Details