Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lys-3
Entrez Gene ID
179430
Gene Name
Lysozyme-like protein 3
Gene Aliases
CELE_Y22F5A.6
Chromosome Location
Chr.V: 10282100 - 10283544 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073243.4 NP_505644.1 1 - 2 450 129

View Full Product Details