Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-53
Entrez Gene ID
179656
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_R07B7.3
Chromosome Location
Chr.V: 12063432 - 12065391 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073622.5 NP_506023.2 1 - 2 151 66
NR_024311.1 1 - 2 119 66

View Full Product Details