Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
che-11
Entrez Gene ID
179666
Gene Name
Uncharacterized protein
Gene Aliases
CELE_C27A7.4
Chromosome Location
Chr.V: 12144244 - 12150707 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073646.7 NP_506047.5 9 - 10 1611 153

View Full Product Details