Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
shw-3
Entrez Gene ID
191761
Gene Name
protein-coding
Gene Aliases
CELE_R186.5
Chromosome Location
Chr.V: 12973261 - 12987161 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368471.3 NP_001355445.1 6 - 7 765 115
NM_001380790.1 NP_001368707.1 6 - 7 756 115
NM_001380791.1 NP_001366842.1 6 - 7 790 115
GenBank mRNA DQ185514.1 - 5 - 6 702 115

View Full Product Details