Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpr-1
Entrez Gene ID
187759
Gene Name
Dauer Pheromone Responsive
Gene Aliases
CELE_R10D12.2
Chromosome Location
Chr.V: 13940139 - 13942182 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074056.4 NP_506457.2 3 - 4 256 95

View Full Product Details