Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-22
Entrez Gene ID
187832
Gene Name
protein-coding
Gene Aliases
CELE_R11H6.3
Chromosome Location
Chr.V: 14600272 - 14606538 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269671.3 NP_001256600.1 5 - 6 765 83
NM_001269672.3 NP_001256601.1 5 - 6 752 83
NM_001269673.4 NP_001256602.1 5 - 6 738 83

View Full Product Details