Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-26
Entrez Gene ID
188144
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T05G11.3
Chromosome Location
Chr.V: 15931292 - 15932490 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322599.1 NP_001309436.1 1 - 2 207 105

View Full Product Details