Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-55
Entrez Gene ID
184963
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F26D2.7
Chromosome Location
Chr.V: 16415694 - 16417114 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074748.1 NP_507149.1 1 - 2 484 146

View Full Product Details