Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-42
Entrez Gene ID
180189
Gene Name
C-type LECtin
Gene Aliases
CELE_F16H6.1
Chromosome Location
Chr.V: 18191412 - 18194168 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075156.4 NP_507557.1 2 - 3 984 75

View Full Product Details