Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scl-21
Entrez Gene ID
189870
Gene Name
SCP domain-containing protein
Gene Aliases
CELE_Y43F8B.5
Chromosome Location
Chr.V: 19494762 - 19495687 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075392.1 NP_507793.1 1 - 2 185 98

View Full Product Details