Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srxa-15
Entrez Gene ID
189895
Gene Name
protein-coding
Gene Aliases
CELE_Y44A6B.2
Chromosome Location
Chr.V: 20642196 - 20644380 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075588.3 NP_507989.2 3 - 4 471 141

View Full Product Details