Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pha-4
Entrez Gene ID
180357
Gene Name
protein-coding
Gene Aliases
CELE_F38A6.1
Chromosome Location
Chr.V: 20753052 - 20760317 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047649.4 NP_001041114.1 1 - 2 45 79

View Full Product Details