Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-21
Entrez Gene ID
180453
Gene Name
CUTiclin-Like
Gene Aliases
CELE_F55A4.10
Chromosome Location
Chr.X: 1004337 - 1009422 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075791.7 NP_508192.2 11 - 12 1300 111

View Full Product Details