Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ttr-37
Entrez Gene ID
184782
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_F21E9.3
Chromosome Location
Chr.X: 1335364 - 1337175 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075844.6 NP_508245.1 2 - 3 293 87

View Full Product Details