Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Y59E1A.4
Entrez Gene ID
42101359
Gene Name
Uncharacterized protein
Gene Aliases
CELE_Y59E1A.4
Chromosome Location
Chr.X: 1939013 - 1939987 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001374972.1 NP_001362147.1 1 - 2 105 79

View Full Product Details